siRNA Details
| Gene Symbol: Cdc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013240.2 | NP_001013258.1 | 4 | 1160 | ||
| NM_001267054.1 | NP_001253983.1 | 4 | 1088 | |||
| NM_001267055.1 | NP_001253984.1 | 3 | 1052 | |||
| XM_006238241.2 | XP_006238303.1 | 4 | 1176 | |||
| XM_006238242.2 | XP_006238304.1 | 3 | 1064 | |||
| XM_008763796.1 | XP_008762018.1 | 4 | 1099 | |||
| GenBank mRNA | BC086343.1 | - | 4 | 1149 | 7301739 | |
| FQ217142.1 | - | 3 | 1043 | |||

Cdc26
Genomic Map