siRNA Details
| Gene Symbol: Tspan13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013244.1 | NP_001013262.1 | 6 | 1814 | ||
| GenBank mRNA | BC089906.1 | - | 6 | 1814 | 7123552 | |
| FQ215999.1 | - | 6 | 1834 | |||
| FQ217050.1 | - | 6 | 1827 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan13| Gene Symbol: Tspan13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013244.1 | NP_001013262.1 | 6 | 1814 | ||
| GenBank mRNA | BC089906.1 | - | 6 | 1814 | 7123552 | |
| FQ215999.1 | - | 6 | 1834 | |||
| FQ217050.1 | - | 6 | 1827 | |||