siRNA Details
Gene Symbol: Sln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013247.1 | NP_001013265.1 | 2 | 119 | ||
GenBank mRNA | AY456000.1 | - | 2 | 119 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013247.1 | NP_001013265.1 | 2 | 119 | ||
GenBank mRNA | AY456000.1 | - | 2 | 119 |