siRNA Details
| Gene Symbol: Dhx30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013249.1 | NP_001013267.1 | 22 | 3784 | ||
| XM_006243826.2 | XP_006243888.1 | 19 | 4911 | |||
| XM_006243827.2 | XP_006243889.1 | 21 | 3672 | |||
| XM_006243829.2 | XP_006243891.1 | 18 | 4800 | |||
| XM_006243830.1 | XP_006243892.1 | 20 | 3994 | |||
| XM_006243831.1 | XP_006243893.1 | 21 | 3640 | |||
| GenBank mRNA | BC091359.1 | - | 22 | 3784 | 7318733 | |

Dhx30
Genomic Map