siRNA Details
| Gene Symbol: Srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013252.1 | NP_001013270.1 | 4 | 414 | ||
| GenBank mRNA | AY325214.1 | - | 2562 | |||
| AY325230.1 | - | 2565 | ||||
| AY325241.1 | - | 2562 | ||||
| BC098951.1 | - | 406 | 7324816 | |||
| BF523222.1 | - | 1 | 80 | |||

Srprb
Genomic Map