siRNA Details
| Gene Symbol: Tcaim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013405.2 | NP_001013423.2 | 5 | 679 | ||
| GenBank mRNA | BC080690.1 | - | 5 | 638 | 30111134 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcaim| Gene Symbol: Tcaim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013405.2 | NP_001013423.2 | 5 | 679 | ||
| GenBank mRNA | BC080690.1 | - | 5 | 638 | 30111134 | |