siRNA Details
| Gene Symbol: LOC100506127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305225.1 | NP_001292154.1 | 1 | 927 | ||
| GenBank mRNA | AK095089.1 | - | 1 | 772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506127| Gene Symbol: LOC100506127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305225.1 | NP_001292154.1 | 1 | 927 | ||
| GenBank mRNA | AK095089.1 | - | 1 | 772 | ||