siRNA Details
| Gene Symbol: PRR23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013650.2 | NP_001013672.1 | 1 | 1193 | ||
| GenBank mRNA | AK127998.1 | - | 1 | 1193 | ||
| BC137145.1 | - | 1 | 1181 | 9021142 | ||
| BC137146.1 | - | 1 | 1179 | 9021143 | ||

PRR23B
Genomic Map