siRNA Details
| Gene Symbol: FRRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013660.2 | NP_001013682.2 | 7 | 1284 | ||
| XM_005270861.3 | XP_005270918.1 | 7 | 811 | |||
| XM_011541451.2 | XP_011539753.1 | 7 | 1284 | |||
| XM_011541452.1 | XP_011539754.1 | 7 | 811 | |||
| XM_017001271.1 | XP_016856760.1 | 5 | 695 | |||
| GenBank mRNA | AK131302.1 | - | 7 | 1284 | ||

FRRS1
Genomic Map