siRNA Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017805.2 | NP_060275.2 | 10 | 2582 | ||
| XM_011527053.2 | XP_011525355.2 | 10 | 2702 | |||
| XM_011527054.2 | XP_011525356.2 | 10 | 2702 | |||
| XM_011527055.2 | XP_011525357.2 | 7 | 2408 | |||
| XM_017026914.1 | XP_016882403.1 | 10 | 2702 | |||
| XM_017026915.1 | XP_016882404.1 | 7 | 2408 | |||
| GenBank mRNA | AK000408.1 | - | 6 | 697 | ||
| AK123179.1 | - | 2 | 879 | |||
| AY378097.1 | - | 10 | 2447 | |||
| BC021860.1 | - | 6 | 712 | |||
| BC028614.1 | - | 10 | 2454 | 4815880 | ||

RASIP1
Genomic Map