siRNA Details
| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013693.2 | NP_001013715.2 | 5 | 3961 | ||
| GenBank mRNA | AK127275.1 | - | 5 | 3961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LDLRAD2| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013693.2 | NP_001013715.2 | 5 | 3961 | ||
| GenBank mRNA | AK127275.1 | - | 5 | 3961 | ||