siRNA Details
| Gene Symbol: IMPAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017813.4 | NP_060283.3 | 2 | 720 | ||
| GenBank mRNA | AK000428.1 | - | 2 | 717 | ||
| AY032885.1 | - | 2 | 434 | |||
| BC017797.1 | - | 2 | 669 | 4306100 | ||
| BC067814.1 | - | 2 | 693 | 5295820 | ||

IMPAD1
Genomic Map