siRNA Details
| Gene Symbol: Aim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013779.2 | NP_001013801.2 | 7 | 1268 | ||
| GenBank mRNA | BC009664.1 | - | 1 | 160 | 3489512 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aim2| Gene Symbol: Aim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013779.2 | NP_001013801.2 | 7 | 1268 | ||
| GenBank mRNA | BC009664.1 | - | 1 | 160 | 3489512 | |