siRNA Details
| Gene Symbol: Fam71b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013783.2 | NP_001013805.1 | 1 | 240 | ||
| GenBank mRNA | AB473727.1 | - | 1 | 165 | ||
| BB615215.1 | - | 1 | 241 | |||
| BC150988.1 | - | 1 | 193 | 9087901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam71b| Gene Symbol: Fam71b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013783.2 | NP_001013805.1 | 1 | 240 | ||
| GenBank mRNA | AB473727.1 | - | 1 | 165 | ||
| BB615215.1 | - | 1 | 241 | |||
| BC150988.1 | - | 1 | 193 | 9087901 | ||