siRNA Details
| Gene Symbol: LOC432459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082906.1 | - | 1 | 1851 | ||
| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143622.1 | - | 2 | 975 | ||
| AK168480.1 | - | 2 | 973 | |||

LOC432459
Genomic Map