siRNA Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013806.1 | NP_001013828.1 | 13 | 2155 | ||
| GenBank mRNA | AK135906.1 | - | 14 | 1891 | ||
| AK157542.1 | - | 2337 | ||||
| BC065088.1 | - | 12 | 4349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd13c| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013806.1 | NP_001013828.1 | 13 | 2155 | ||
| GenBank mRNA | AK135906.1 | - | 14 | 1891 | ||
| AK157542.1 | - | 2337 | ||||
| BC065088.1 | - | 12 | 4349 | |||