siRNA Details
| Gene Symbol: Fam169b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013811.2 | NP_001013833.1 | 9 | 2800 | ||
| GenBank mRNA | BC039209.1 | - | 9 | 2828 | 4161789 | |
| BC039945.1 | - | 9 | 2839 | 4161789 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam169b| Gene Symbol: Fam169b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013811.2 | NP_001013833.1 | 9 | 2800 | ||
| GenBank mRNA | BC039209.1 | - | 9 | 2828 | 4161789 | |
| BC039945.1 | - | 9 | 2839 | 4161789 | ||