siRNA Details
| Gene Symbol: Krtap4-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013823.1 | NP_001013845.1 | 1 | 732 | ||
| GenBank mRNA | BC016249.1 | - | 1 | 732 | 4910141 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap4-16| Gene Symbol: Krtap4-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013823.1 | NP_001013845.1 | 1 | 732 | ||
| GenBank mRNA | BC016249.1 | - | 1 | 732 | 4910141 | |