siRNA Details
| Gene Symbol: LOC297756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013928.2 | NP_001013950.2 | 1 | 46 | ||
| GenBank mRNA | AY383664.1 | - | 1 | 46 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC297756| Gene Symbol: LOC297756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013928.2 | NP_001013950.2 | 1 | 46 | ||
| GenBank mRNA | AY383664.1 | - | 1 | 46 | ||