siRNA Details
| Gene Symbol: IgG-2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088240.1 | - | 5 | 723 | 7365825 | |
| BC088423.1 | - | 5 | 795 | 7123713 | ||
| BC091257.1 | - | 724 | 7375893 | |||
| BC091272.1 | - | 726 | 7382505 | |||
| L22652.1 | - | 6 | 792 | |||
| L22654.1 | - | 806 | ||||

IgG-2a
Genomic Map