siRNA Details
| Gene Symbol: IgG-2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088240.1 | - | 2 | 159 | 7365825 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

IgG-2a| Gene Symbol: IgG-2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088240.1 | - | 2 | 159 | 7365825 | |