siRNA Details
| Gene Symbol: Slain1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014139.1 | NP_001014161.1 | 5 | 1094 | ||
| GenBank mRNA | BC088867.1 | - | 5 | 1094 | 7320965 | |
| FQ226954.1 | - | 1 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slain1| Gene Symbol: Slain1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014139.1 | NP_001014161.1 | 5 | 1094 | ||
| GenBank mRNA | BC088867.1 | - | 5 | 1094 | 7320965 | |
| FQ226954.1 | - | 1 | 142 | |||