siRNA Details
| Gene Symbol: Tmem192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014141.1 | NP_001014163.1 | 5 | 789 | ||
| GenBank mRNA | BC086402.1 | - | 5 | 789 | 7304126 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem192| Gene Symbol: Tmem192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014141.1 | NP_001014163.1 | 5 | 789 | ||
| GenBank mRNA | BC086402.1 | - | 5 | 789 | 7304126 | |