siRNA Details
| Gene Symbol: Ufsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014142.1 | NP_001014164.1 | 9 | 1063 | ||
| XM_006253159.2 | XP_006253221.1 | 9 | 1053 | |||
| XM_008771265.1 | XP_008769487.1 | 9 | 1214 | |||
| GenBank mRNA | BC083771.1 | - | 9 | 1063 | 7193395 | |

Ufsp2
Genomic Map