siRNA Details
| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014145.1 | NP_001014167.1 | 5, 6 | 1085 | ||
| GenBank mRNA | BC079003.1 | - | 5, 6 | 1085 | 7112869 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdyl| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014145.1 | NP_001014167.1 | 5, 6 | 1085 | ||
| GenBank mRNA | BC079003.1 | - | 5, 6 | 1085 | 7112869 | |