siRNA Details
| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014145.1 | NP_001014167.1 | 6 | 1135 | ||
| XM_006253839.2 | XP_006253901.1 | 4 | 1325 | |||
| XM_006253840.2 | XP_006253902.1 | 4 | 1185 | |||
| GenBank mRNA | BC079003.1 | - | 6 | 1135 | 7112869 | |

Cdyl
Genomic Map