siRNA Details
| Gene Symbol: Paip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014148.1 | NP_001014170.1 | 6 | 1545 | ||
| XM_008772044.1 | XP_008770266.1 | 4 | 1392 | |||
| XM_008772046.1 | XP_008770268.1 | 4 | 1426 | |||
| XM_008772047.1 | XP_008770269.1 | 5 | 1448 | |||
| XM_008772048.1 | XP_008770270.1 | 5 | 1465 | |||
| XM_008772049.1 | XP_008770271.1 | 4 | 1538 | |||
| GenBank mRNA | BC079261.1 | - | 6 | 1545 | 7133501 | |

Paip2
Genomic Map