siRNA Details
| Gene Symbol: Rell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014149.1 | NP_001014171.1 | 8 | 1634 | ||
| GenBank mRNA | BC089946.1 | - | 8 | 1634 | 7311493 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rell2| Gene Symbol: Rell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014149.1 | NP_001014171.1 | 8 | 1634 | ||
| GenBank mRNA | BC089946.1 | - | 8 | 1634 | 7311493 | |