siRNA Details
Gene Symbol: Yipf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254664.2 | XP_006254726.1 | 2 | 80 | ||
GenBank mRNA | AY321347.1 | - | 2 | 86 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Yipf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254664.2 | XP_006254726.1 | 2 | 80 | ||
GenBank mRNA | AY321347.1 | - | 2 | 86 |