siRNA Details
| Gene Symbol: Hdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254965.2 | XP_006255027.1 | 7 | 1038 | ||
| XM_006254966.2 | XP_006255028.1 | 7 | 1491 | |||
| GenBank mRNA | BC078941.1 | - | 7 | 1012 | 7109225 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdhd2| Gene Symbol: Hdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254965.2 | XP_006255027.1 | 7 | 1038 | ||
| XM_006254966.2 | XP_006255028.1 | 7 | 1491 | |||
| GenBank mRNA | BC078941.1 | - | 7 | 1012 | 7109225 | |