siRNA Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014164.1 | NP_001014186.1 | 6 | 1738 | ||
| XM_006231049.2 | XP_006231111.1 | 7 | 1837 | |||
| GenBank mRNA | BC085835.1 | - | 6 | 1738 | 7134938 | |
| FQ234846.1 | - | 1 | 28 | |||

Cyb561a3
Genomic Map