siRNA Details
| Gene Symbol: Ccdc172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014169.2 | NP_001014191.1 | 9 | 872 | ||
| XM_008760541.1 | XP_008758763.1 | 8 | 1161 | |||
| XM_008760542.1 | XP_008758764.1 | 8 | 1158 | |||
| GenBank mRNA | BC079324.1 | - | 8 | 1089 | 7135457 | |

Ccdc172
Genomic Map