siRNA Details
| Gene Symbol: Ccdc172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014169.2 | NP_001014191.1 | 4 | 326 | ||
| XM_006231674.2 | XP_006231736.1 | 4 | 342 | |||
| XM_008760539.1 | XP_008758761.1 | 3 | 615 | |||
| XM_008760541.1 | XP_008758763.1 | 3 | 615 | |||
| XM_008760542.1 | XP_008758764.1 | 3 | 615 | |||
| GenBank mRNA | BC079324.1 | - | 3 | 543 | 7135457 | |
| CV104909.1 | - | 4 | 326 | |||

Ccdc172
Genomic Map