siRNA Details
| Gene Symbol: Alg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014176.2 | NP_001014198.1 | 4 | 696 | ||
| XM_008761465.1 | XP_008759687.1 | 5 | 868 | |||
| GenBank mRNA | BC079151.1 | - | 4 | 642 | 7128610 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg14| Gene Symbol: Alg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014176.2 | NP_001014198.1 | 4 | 696 | ||
| XM_008761465.1 | XP_008759687.1 | 5 | 868 | |||
| GenBank mRNA | BC079151.1 | - | 4 | 642 | 7128610 | |