siRNA Details
| Gene Symbol: Spata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014177.1 | NP_001014199.1 | 11 | 1318 | ||
| XM_008761517.1 | XP_008759739.1 | 11 | 1607 | |||
| GenBank mRNA | BC079224.1 | - | 11 | 1318 | 7132232 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata1| Gene Symbol: Spata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014177.1 | NP_001014199.1 | 11 | 1318 | ||
| XM_008761517.1 | XP_008759739.1 | 11 | 1607 | |||
| GenBank mRNA | BC079224.1 | - | 11 | 1318 | 7132232 | |