siRNA Details
| Gene Symbol: Fam69b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014178.2 | NP_001014200.2 | 5 | 979 | ||
| GenBank mRNA | BC089911.1 | - | 1 | 548 | ||
| EV771144.1 | - | 3 | 659 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam69b| Gene Symbol: Fam69b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014178.2 | NP_001014200.2 | 5 | 979 | ||
| GenBank mRNA | BC089911.1 | - | 1 | 548 | ||
| EV771144.1 | - | 3 | 659 | |||