siRNA Details
| Gene Symbol: Lancl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014187.1 | NP_001014209.1 | 7 | 1184 | ||
| XM_006236580.2 | XP_006236642.2 | 7 | 1046 | |||
| XM_006236581.2 | XP_006236643.1 | 7 | 1193 | |||
| GenBank mRNA | BC079412.1 | - | 7 | 1184 | 7113320 | |

Lancl2
Genomic Map