siRNA Details
| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC085888.1 | - | 3 | 1792 | 7192402 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsrp1| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC085888.1 | - | 3 | 1792 | 7192402 | |