siRNA Details
| Gene Symbol: Tmem214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014195.2 | NP_001014217.2 | 12 | 1417 | ||
| GenBank mRNA | BC085114.1 | - | 9 | 848 | 7106107 | |
| BC129115.1 | - | 12 | 1417 | 7112909 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem214| Gene Symbol: Tmem214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014195.2 | NP_001014217.2 | 12 | 1417 | ||
| GenBank mRNA | BC085114.1 | - | 9 | 848 | 7106107 | |
| BC129115.1 | - | 12 | 1417 | 7112909 | ||