siRNA Details
| Gene Symbol: BCAS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010974.2 | NP_001010974.1 | 3 | 390 | ||
| NM_017843.4 | NP_060313.3 | 3 | 390 | |||
| NM_198799.3 | NP_942094.2 | 3 | 390 | |||
| XM_011528886.2 | XP_011527188.1 | 3 | 313 | |||
| XM_011528887.2 | XP_011527189.1 | 3 | 313 | |||
| XM_017027932.1 | XP_016883421.1 | 3 | 313 | |||
| GenBank mRNA | AF361220.1 | - | 3 | 390 | ||
| AJ511266.1 | - | 3 | 214 | |||
| AK000502.1 | - | 3 | 253 | |||
| BC038381.1 | - | 3 | 240 | 4472674 | ||
| BC047337.1 | - | 3 | 219 | |||
| BP212127.1 | - | 3 | 354 | |||

BCAS4
Genomic Map