siRNA Details
| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014198.2 | NP_001014220.2 | 3 | 904 | ||
| XM_006240527.2 | XP_006240589.1 | 3 | 1275 | |||
| GenBank mRNA | BC087681.1 | - | 3 | 881 | 7321205 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gskip| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014198.2 | NP_001014220.2 | 3 | 904 | ||
| XM_006240527.2 | XP_006240589.1 | 3 | 1275 | |||
| GenBank mRNA | BC087681.1 | - | 3 | 881 | 7321205 | |