siRNA Details
| Gene Symbol: Atp6v1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014199.1 | NP_001014221.1 | 14 | 1440 | ||
| GenBank mRNA | AY594319.1 | - | 13 | 1365 | ||
| BC079083.1 | - | 14 | 1440 | 7122992 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1c2| Gene Symbol: Atp6v1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014199.1 | NP_001014221.1 | 14 | 1440 | ||
| GenBank mRNA | AY594319.1 | - | 13 | 1365 | ||
| BC079083.1 | - | 14 | 1440 | 7122992 | ||