siRNA Details
| Gene Symbol: Tmem123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014205.1 | NP_001014227.1 | 4 | 1412 | ||
| GenBank mRNA | BC089103.1 | - | 4 | 1412 | 7307876 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem123| Gene Symbol: Tmem123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014205.1 | NP_001014227.1 | 4 | 1412 | ||
| GenBank mRNA | BC089103.1 | - | 4 | 1412 | 7307876 | |