siRNA Details
| Gene Symbol: RGD1309534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014206.1 | NP_001014228.1 | 9 | 1321 | ||
| GenBank mRNA | BC085915.1 | - | 9 | 1321 | 7107444 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309534| Gene Symbol: RGD1309534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014206.1 | NP_001014228.1 | 9 | 1321 | ||
| GenBank mRNA | BC085915.1 | - | 9 | 1321 | 7107444 | |