siRNA Details
| Gene Symbol: Taf1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014207.1 | NP_001014229.1 | 6 | 882 | ||
| XM_008765945.1 | XP_008764167.1 | 6 | 1799 | |||
| XM_008765946.1 | XP_008764168.1 | 6 | 1799 | |||
| XM_008765947.1 | XP_008764169.1 | 6 | 1799 | |||
| XM_008765948.1 | XP_008764170.1 | 6 | 1799 | |||
| GenBank mRNA | BC087647.1 | - | 6 | 882 | 7311303 | |

Taf1d
Genomic Map