siRNA Details
| Gene Symbol: Taf1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014207.1 | NP_001014229.1 | 9 | 1003 | ||
| XM_008765941.1 | XP_008764163.1 | 9 | 2589 | |||
| XM_008765942.1 | XP_008764164.1 | 10 | 1752 | |||
| XM_008765943.1 | XP_008764165.1 | 10 | 2044 | |||
| XM_008765945.1 | XP_008764167.1 | 9 | 3111 | |||
| XM_008765946.1 | XP_008764168.1 | 10 | 1965 | |||
| XM_008765947.1 | XP_008764169.1 | 9 | 1920 | |||
| GenBank mRNA | BC087647.1 | - | 9 | 1003 | 7311303 | |

Taf1d
Genomic Map