siRNA Details
| Gene Symbol: Plet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014209.1 | NP_001014231.1 | 1 | 182 | ||
| GenBank mRNA | BC088858.1 | - | 1 | 182 | 7308076 | |
| FQ228964.1 | - | 1 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plet1| Gene Symbol: Plet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014209.1 | NP_001014231.1 | 1 | 182 | ||
| GenBank mRNA | BC088858.1 | - | 1 | 182 | 7308076 | |
| FQ228964.1 | - | 1 | 205 | |||