siRNA Details
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014215.1 | NP_001014237.1 | 2 | 253 | ||
GenBank mRNA | BC082113.1 | - | 2 | 253 | 7135459 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014215.1 | NP_001014237.1 | 2 | 253 | ||
GenBank mRNA | BC082113.1 | - | 2 | 253 | 7135459 |