siRNA Details
| Gene Symbol: Sp100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014220.1 | NP_001014242.1 | 5 | 675 | ||
| GenBank mRNA | BC083808.1 | - | 5 | 675 | 7109918 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp100| Gene Symbol: Sp100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014220.1 | NP_001014242.1 | 5 | 675 | ||
| GenBank mRNA | BC083808.1 | - | 5 | 675 | 7109918 | |