siRNA Details
| Gene Symbol: Dmrtc1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014222.2 | NP_001014244.2 | 9 | 1178 | ||
| GenBank mRNA | BC079417.1 | - | 9 | 1164 | 7113562 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmrtc1c1| Gene Symbol: Dmrtc1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014222.2 | NP_001014244.2 | 9 | 1178 | ||
| GenBank mRNA | BC079417.1 | - | 9 | 1164 | 7113562 | |